Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl 1,10-decanedicarboxylate, 98%

$86.19 - $320.95

Chemical Identifiers

Chemical Name or Material Dimethyl 1,10-decanedicarboxylate
CAS 1731-79-9
Molecular Formula C14H26O4
Linear Formula CH3O2C(CH2)10CO2CH3
Formula Weight 258.36
MDL Number MFCD00043632
Beilstein 1790424
Synonym 1,10-Decanedicarboxylic acid dimethyl ester; Dodecanedioic acid dimethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0875718

  SDS
alfa aesar™
L0875718
50g Each for $320.95

AAL0875709

  SDS
alfa aesar™
L0875709
10g Each for $86.19
Description & Specifications

Specifications

Chemical Name or Material Dimethyl 1,10-decanedicarboxylate
CAS 1731-79-9
TSCA TSCA
Assay 98%
Molecular Formula C14H26O4
Melting Point 27° to 32°C
Linear Formula CH3O2C(CH2)10CO2CH3
Boiling Point 187° to 188°C (14mmHg)
Formula Weight 258.36
MDL Number MFCD00043632
Beilstein 1790424
Quantity 50g
Synonym 1,10-Decanedicarboxylic acid dimethyl ester; Dodecanedioic acid dimethyl ester