Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethoxymethylsilane, 97%

Chemical Identifiers

Chemical Name or Material Dimethoxymethylsilane
CAS 16881-77-9
Molecular Formula C3H10O2Si
Formula Weight 106.2
MDL Number MFCD00048019
UN Number UN1993
Beilstein 2035898
Synonym Methyldimethoxysilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1619706

  SDS
alfa aesar™
L1619706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1619714

  SDS
alfa aesar™
L1619714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 106.2
Molecular Formula C3H10O2Si
MDL Number MFCD00048019
Synonym Methyldimethoxysilane
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Dimethoxymethylsilane
TSCA TSCA
Assay 97%
CAS 16881-77-9
UN Number UN1993
Melting Point −136°C
Flash Point 2°C (35°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H318-H335-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Beilstein 2035898
Boiling Point 61°C
Quantity 5g
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H318-H335-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.