Please login to your online account to display your discounted pricing

Dimethoxydimethylsilane, 95+%, AcroSeal™, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Dimethoxydimethylsilane
Assay ≥94%
Assay Percent Range 95+%
Molecular Formula C4H12O2Si
Linear Formula (CH3)Si(OCH3)
Formula Weight 120.23
Synonym Dimethyldimethoxysilane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC351530010

  SDS
thermo fisher scientific
AC351530010
AcroSeal™ Glass Bottle 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC351531000

  SDS
thermo fisher scientific
AC351531000
AcroSeal™ Glass Bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 120.23
Assay ≥94%
Physical Form Liquid
Quantity 1L
Color Clear colorless to light yellow
Packaging AcroSeal™ Glass Bottle
Density 0.865g/mL
Synonym Dimethyldimethoxysilane
Refractive Index 1.3690 to 1.3710 (20°C, 589nm)
Health Hazard 1 Danger
Chemical Name or Material Dimethoxydimethylsilane
Assay Percent Range 95+%
Molecular Formula C4H12O2Si
CAS 1112-39-6
Infrared Spectrum Authentic
Linear Formula (CH3)Si(OCH3)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Safety and Handling

Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.