Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethallyl carbonate, 97%

$55.89 - $55.89

Chemical Identifiers

Chemical Name or Material Dimethallyl carbonate
CAS 64057-79-0
Molecular Formula C11H18O3
Formula Weight 170.21
MDL Number MFCD00008598
Synonym Bis(2-methylallyl) carbonate; Carbonic acid dimethallyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2041406

  SDS
alfa aesar™
B2041406
5g Each for $55.89
Description & Specifications

Specifications

Formula Weight 170.21
Boiling Point 199° to 202°C
Synonym Bis(2-methylallyl) carbonate; Carbonic acid dimethallyl ester
Chemical Name or Material Dimethallyl carbonate
TSCA TSCA
Assay 97%
Molecular Formula C11H18O3
MDL Number MFCD00008598
CAS 64057-79-0
Quantity 5g
RTECS Number FF8750000

Applications
It is used as an intermediate in enantioselective tsuji enol silane allylation reaction.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.