missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimesitylmethane, 98+%

$30.84 - $115.97

Chemical Identifiers

CAS 733-07-3
Molecular Formula C19H24
Molecular Weight (g/mol) 252.401
MDL Number MFCD00014913
InChI Key ZQYQPPLKPFBKLD-UHFFFAOYSA-N
Synonym dimesitylmethane, 1,1'-methylenebis 2,4,6-trimethylbenzene, 1,3,5-trimethyl-2-2,4,6-trimethylphenyl methyl benzene, benzene, 1,1'-methylenebis 2,4,6-trimethyl, benzene,1'-methylenebis 2,4,6-trimethyl, dimesitylmethane, 2-mesitylmethyl-1,3,5-trimethylbenzene, 2-mesitylmethyl-1,3,5-trimethylbenzene #, benzene,1,3,5-trimethyl-2-2,4,6-trimethylphenyl methyl
PubChem CID 69771
IUPAC Name 1,3,5-trimethyl-2-[(2,4,6-trimethylphenyl)methyl]benzene
SMILES CC1=CC(=C(C(=C1)C)CC2=C(C=C(C=C2C)C)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0109006 View Documents Alfa Aesar™
L0109006
5g Each for $30.84
 
AAL0109014 View Documents Alfa Aesar™
L0109014
25g Each for $115.97
 
Specifications

Chemical Identifiers

733-07-3
252.401
ZQYQPPLKPFBKLD-UHFFFAOYSA-N
69771
CC1=CC(=C(C(=C1)C)CC2=C(C=C(C=C2C)C)C)C
C19H24
MFCD00014913
dimesitylmethane, 1,1'-methylenebis 2,4,6-trimethylbenzene, 1,3,5-trimethyl-2-2,4,6-trimethylphenyl methyl benzene, benzene, 1,1'-methylenebis 2,4,6-trimethyl, benzene,1'-methylenebis 2,4,6-trimethyl, dimesitylmethane, 2-mesitylmethyl-1,3,5-trimethylbenzene, 2-mesitylmethyl-1,3,5-trimethylbenzene #, benzene,1,3,5-trimethyl-2-2,4,6-trimethylphenyl methyl
1,3,5-trimethyl-2-[(2,4,6-trimethylphenyl)methyl]benzene

Specifications

129°C to 131°C
733-07-3
MFCD00014913
dimesitylmethane, 1,1'-methylenebis 2,4,6-trimethylbenzene, 1,3,5-trimethyl-2-2,4,6-trimethylphenyl methyl benzene, benzene, 1,1'-methylenebis 2,4,6-trimethyl, benzene,1'-methylenebis 2,4,6-trimethyl, dimesitylmethane, 2-mesitylmethyl-1,3,5-trimethylbenzene, 2-mesitylmethyl-1,3,5-trimethylbenzene #, benzene,1,3,5-trimethyl-2-2,4,6-trimethylphenyl methyl
CC1=CC(=C(C(=C1)C)CC2=C(C=C(C=C2C)C)C)C
252.401
252.4
Dimesitylmethane
5g
C19H24
2053472
ZQYQPPLKPFBKLD-UHFFFAOYSA-N
1,3,5-trimethyl-2-[(2,4,6-trimethylphenyl)methyl]benzene
69771
≥98%