missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimesitylborinic acid, 98%

$32.95 - $437.16

Chemical Identifiers

CAS 20631-84-9
Molecular Formula C18H23BO
Molecular Weight (g/mol) 266.191
MDL Number MFCD01863507
InChI Key OTYVTANMHIUXBQ-UHFFFAOYSA-N
Synonym bis 2,4,6-trimethylphenyl borinic acid
PubChem CID 5102928
IUPAC Name bis(2,4,6-trimethylphenyl)borinic acid
SMILES B(C1=C(C=C(C=C1C)C)C)(C2=C(C=C(C=C2C)C)C)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5545503 View Documents Alfa Aesar™
H5545503
1g Each for $32.95
 
AAH5545506 View Documents Alfa Aesar™
H5545506
5g Each for $117.00
 
AAH5545514 View Documents Alfa Aesar™
H5545514
25g Each for $437.16
 
Specifications

Chemical Identifiers

20631-84-9
266.191
OTYVTANMHIUXBQ-UHFFFAOYSA-N
5102928
B(C1=C(C=C(C=C1C)C)C)(C2=C(C=C(C=C2C)C)C)O
C18H23BO
MFCD01863507
bis 2,4,6-trimethylphenyl borinic acid
bis(2,4,6-trimethylphenyl)borinic acid

Specifications

144°C to 146°C
20631-84-9
MFCD01863507
OTYVTANMHIUXBQ-UHFFFAOYSA-N
bis(2,4,6-trimethylphenyl)borinic acid
5102928
Solid
Dimesitylborinic acid
1g
C18H23BO
bis 2,4,6-trimethylphenyl borinic acid
B(C1=C(C=C(C=C1C)C)C)(C2=C(C=C(C=C2C)C)C)O
266.191
266.19
98%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures