missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimesitylborinic acid, 98%

Manufacturer:  Alfa Aesar™ H5545514

 View more versions of this product

Catalog No. AAH5545514



Specifications

Specifications

Dimesitylborinic acid
25g
C18H23BO
bis 2,4,6-trimethylphenyl borinic acid
B(C1=C(C=C(C=C1C)C)C)(C2=C(C=C(C=C2C)C)C)O
266.191
266.19
98%
144°C to 146°C
20631-84-9
MFCD01863507
OTYVTANMHIUXBQ-UHFFFAOYSA-N
bis(2,4,6-trimethylphenyl)borinic acid
5102928
Solid

Chemical Identifiers

20631-84-9
266.191
OTYVTANMHIUXBQ-UHFFFAOYSA-N
5102928
B(C1=C(C=C(C=C1C)C)C)(C2=C(C=C(C=C2C)C)C)O
C18H23BO
MFCD01863507
bis 2,4,6-trimethylphenyl borinic acid
bis(2,4,6-trimethylphenyl)borinic acid
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents