missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-Dimenthyl fumarate, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC429680010

 View more versions of this product

Catalog No. AC429680010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94.0%
Low Melting Solid or Powder
Colorless to off-white
100.0
C24H40O4
95%
Authentic
392.58
1g
94.0
-95° to -105° (20°C, 589nm) (C=1 in chloroform)
34675-24-6
(-)-Dimenthyl fumarate

Chemical Identifiers

34675-24-6
100.0
C24H40O4
(-)-Dimenthyl fumarate
94.0
95%
392.58
≥94.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.