missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-Dimenthyl fumarate, 95%, ACROS Organics™

Chemical Identifiers

CAS 34675-24-6
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C24H40O4
Formula Weight 392.58
Chemical Name or Material (-)-Dimenthyl fumarate
Assay ≥94.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC429680050 SDS Thermo Fisher Scientific
AC429680050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC429680010 SDS Thermo Fisher Scientific
AC429680010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

34675-24-6
100.0
C24H40O4
(-)-Dimenthyl fumarate
94.0
95%
392.58
≥94.0%

Specifications

392.58
5g
94.0
-95° to -105° (20°C, 589nm) (C=1 in chloroform)
C24H40O4
Authentic
≥94.0%
Low Melting Solid or Powder
Colorless to off-white
100.0
34675-24-6
95%
(-)-Dimenthyl fumarate

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.