missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimenhydrinate

A histamine H1 antagonist

Manufacturer:  Alfa Aesar™ J6371806

 View more versions of this product

Catalog No. AAJ6371806



Description

Description

Solubility
Soluble in ethanol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

Dimenhydrinate
102°C to 107°C
523-87-5
MFCD00054265
amosyt, anautine, aviomarin, chloranautine, dimenhydrinate, diphenhydrinate, dramamine, gravol, travelin, vomex a
NFLLKCVHYJRNRH-UHFFFAOYSA-N
2-benzhydryloxy-N,N-dimethylethanamine;8-chloro-1,3-dimethyl-7H-purine-2,6-dione
10660
Powder
White
5g
C24H28ClN5O3
14,3207
Soluble in ethanol
CN1C2=C(C(=O)N(C1=O)C)NC(=N2)Cl.CN(C)CCOC(C1=CC=CC=C1)C2=CC=CC=C2
469.97
469.96

Chemical Identifiers

523-87-5
469.97
NFLLKCVHYJRNRH-UHFFFAOYSA-N
10660
CN1C2=C(C(=O)N(C1=O)C)NC(=N2)Cl.CN(C)CCOC(C1=CC=CC=C1)C2=CC=CC=C2
C24H28ClN5O3
MFCD00054265
amosyt, anautine, aviomarin, chloranautine, dimenhydrinate, diphenhydrinate, dramamine, gravol, travelin, vomex a
2-benzhydryloxy-N,N-dimethylethanamine;8-chloro-1,3-dimethyl-7H-purine-2,6-dione
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XH5082000

TSCA : TSCA

Documents