missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimedone, 98%

Manufacturer:  Alfa Aesar™ A1014036

 View more versions of this product

Catalog No. AAA1014036



Description

Description

Applications
Reagent used in synthesis of fused 3-methylfuransDimedone is used in organic chemistry to identify the presence of aldehyde group. It is also used as a catalyst in the formation of transition-metal complexes. Further, it is also employed in colorimetry, crystallography, luminescence and spectrophotometric analysis. In addition to this, it is used in the synthesis of naturally occurring fused 3-methylfurans.

Solubility
Soluble in methanol, ethanol, benzene and chloroform. Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents, strong acids and strong bases.
Specifications

Specifications

Dimedone
500g
C8H12O2
471489
dimedone, 5,5-dimethyl-1,3-cyclohexanedione, cyclomethone, medon, methone, dimedon, methon, 1,3-cyclohexanedione, 5,5-dimethyl, 5,5-dimethyldihydroresorcinol, 5,5-dimethylhydroresorcinol
CC1(CC(=O)CC(=O)C1)C
140.182
140.18
146°C to 150°C
126-81-8
MFCD00001588
14,3242
BADXJIPKFRBFOT-UHFFFAOYSA-N
5,5-dimethylcyclohexane-1,3-dione
31358
98%

Chemical Identifiers

126-81-8
140.182
BADXJIPKFRBFOT-UHFFFAOYSA-N
31358
CC1(CC(=O)CC(=O)C1)C
C8H12O2
MFCD00001588
dimedone, 5,5-dimethyl-1,3-cyclohexanedione, cyclomethone, medon, methone, dimedon, methon, 1,3-cyclohexanedione, 5,5-dimethyl, 5,5-dimethyldihydroresorcinol, 5,5-dimethylhydroresorcinol
5,5-dimethylcyclohexane-1,3-dione
Safety and Handling

Safety and Handling

RTECSNumber : GV0345000

TSCA : TSCA

SDS Documents