Please login to your online account to display your discounted pricing

Dimebon dihydrochloride, Tocris

Neuroprotectant; protects against β-amyloid neurotoxicity

$133.98 - $284.81

Chemical Identifiers

Chemical Name or Material Dimebon dihydrochloride
Assay Percent Range >98%
Molecular Formula C21H25N3·2HCl
Formula Weight 392.37
Synonym 2,3,4,5-Tetrahydro-2,8-dimethyl-5-[2-(6-methyl-3-pyridinyl)ethyl]-1H-pyrido[4,3-b]indole dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

32011

 
r & d systems
32011
1mg Each for $133.98

320110

 
r & d systems
320110
10mg Each for $284.81
Description & Specifications

Specifications

Formula Weight 392.37
Quantity 1mg
CAS 97657-92-6
Molecular Formula C21H25N3·2HCl
Synonym 2,3,4,5-Tetrahydro-2,8-dimethyl-5-[2-(6-methyl-3-pyridinyl)ethyl]-1H-pyrido[4,3-b]indole dihydrochloride
Assay Percent Range >98%
Chemical Name or Material Dimebon dihydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature