Dimco-Gray GraLab 170 Series Timers
Offers precise timing for industrial and laboratory work—split-second accuracy
Manufacturer:  Dimco-Gray DIM171CSA
 View more versions of this product
Catalog No. 06-656
Description
Description
Offers precise timing for industrial and laboratory work—split-second accuracy
Accuracy even at maximum setting, plus ease-of-use features like large 6.5 in. face.
  • Two switchable 3-wire receptacles: control one appliance with a switch that can either make or break circuit at zero; other receptacle is independent of switch and always breaks at zero
  • Outer ring shows elapsed time; inner can be used as stop clock; red numerals count up, black numerals count down
  • Power switch starts timing cycle
  • Rubber base pads, wall mounting keyholes and recessed hand grip offers multiple placement options
  • Study 7.5 × 7.5 × 2.5 in. polycarbonate/PET case
  • Permanent boot over power switch to prevent contamination and corrosion
06-656-1C (Model 170):
“Minutes” hand rotates one revolution/60 min.
“Hours” hand rotates one revolution/60 hr.
06-656 (Model 171):
“Seconds” hand rotates one revolution/60 sec.
“Minutes” hand rotates one revolution/60 min.
06-656-1G (Model 172):
“Seconds” hand rotates one revolution/15 sec.
“Minutes” hand rotates one revolution/15 min.
06-656-1H (Model 173):
“Minutes” hand rotates one revolution/15 min.
“Hours” hand rotates one revolution/15 hr.
Warranty
Limited one year
Specifications
Specifications
UL, cUL
50/60 Hz
Outer ring, Rubber base pads, wall mounting keyholes and recessed hand grip, study 7.5 x 7.5 x 2.5 in. Polycarbonate/PET case, permanent boot over power switch
100/130 VAC
Analog Stopclock
100/130 VAC 50/60 Hz
0.33
Two switchable 3-wire receptacles
±0.15%
0 to 60 min.