Please login to your online account to display your discounted pricing

Dimanganese decacarbonyl, 98%, ACROS Organics™

$56.32 - $56.32

Chemical Identifiers

Chemical Name or Material Dimanganese decacarbonyl
Assay Percent Range 98%
Molecular Formula C10Mn2O10
Linear Formula Mn(CO)10
Formula Weight 389.98
MDL Number MFCD00011115
Synonym Manganese carbonyl
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC315310010

  SDS
thermo fisher scientific
315310010
Glass bottle 1g Each for $56.32
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 389.98
Quantity 1g
Color Yellow
Merck Index 15, 5792
Infrared Spectrum Authentic
Linear Formula Mn(CO)10
Molecular Formula C10Mn2O10
Assay Percent Range 98%
CAS 10170-69-1
Chemical Name or Material Dimanganese decacarbonyl
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00011115
Packaging Glass bottle
Elemental Analysis 97.5% min.
Synonym Manganese carbonyl
Health Hazard 3 GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use only outdoors or in a well-ventilated area.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use only outdoors or in a well-ventilated area.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.