missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimanganese decacarbonyl, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 315310010

 View more versions of this product

Catalog No. AC315310010


 

Specifications Description & Specifications

Specifications

Crystalline Powder
1g
15, 5792
Mn(CO)10
98%
Dimanganese decacarbonyl
Glass bottle
Manganese carbonyl
389.98
Yellow
Authentic
C10Mn2O10
10170-69-1
MFCD00011115
97.5% min.

Chemical Identifiers

98%
Mn(CO)10
MFCD00011115
Dimanganese decacarbonyl
C10Mn2O10
389.98
Manganese carbonyl

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.

GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use only outdoors or in a well-ventilated area.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger