Please login to your online account to display your discounted pricing

Diluting Fluid for CSF (Cerebrospinal Fluid), LabChem™

$47.37 - $47.37

Chemical Identifiers

Chemical Name or Material Diluting Fluid for CSF (Cerebrospinal Fluid)
CAS 7732-18-5
CAS Max % 89.92
CAS 64-19-7
CAS Max % 9.88
CAS 548-62-9
CAS Max % 0.2
Linear Formula C25H30CIN3
Formula Weight 407.99
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC135857

 
lab chem
LC135857
125mL Each for $47.37
Description & Specifications

Specifications

Formula Weight 407.99
Chemical Name or Material Diluting Fluid for CSF (Cerebrospinal Fluid)
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Color Dark purple
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 9.88
Specific Gravity 1.01g/mL
Storage Information Room Temperature
Viscosity 1.2cSt
CAS Max % 0.2
Quantity 125mL
Physical Form Liquid
Linear Formula C25H30CIN3
Density 1.01g/mL
CAS 7732-18-5
Health Hazard 1 Warning
CAS Max % 89.92
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing cancer.
Harmful to aquatic life.
Identification Passes Test
CAS 64-19-7
Packaging Poly Bottle
Solubility Soluble in water
CAS 548-62-9
Safety and Handling

Warning

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing cancer.
Harmful to aquatic life.