Please login to your online account to display your discounted pricing

Dilu-Lok™ Dilution Vials


Designed for testing in the food, dairy, pharmaceutical, water, and cosmetic industries

Manufacturer: hardy diagnostics  D699

 View more versions of this product

Catalog No. 02-686-202B

  • / Case of 50 Each
 

Description & Specifications

Specifications

Volume (Metric) 99mL, Phosphate Buffer w/ Magnesium Chloride
Capacity 3.34 oz., 99mL
Certifications/Compliance APHA, AOAC, FDA, USDA
Closure Type Hinged
Quantity 50 Case
Volume (English) 3.34 oz.
Capacity (English) 3.34 oz.
Capacity (Metric) 99mL
For Use With (Application) Designed for testing in the food, dairy, pharmaceutical, water, and cosmetic industries
Material Polypropylene