Please login to your online account to display your discounted pricing

Dilu-Lok™ Dilution Vials


Designed for testing in the food, dairy, pharmaceutical, water, and cosmetic industries

Manufacturer: hardy diagnostics  D690

 View more versions of this product

Catalog No. 02-686-202A

  • / Case of 50 Each
 

Description & Specifications

Specifications

Volume (Metric) 90mL, Phosphate Buffer w/ Magnesium Chloride
For Use With (Application) Designed for testing in the food, dairy, pharmaceutical, water, and cosmetic industries
Capacity (Metric) 90mL
Certifications/Compliance APHA, AOAC, FDA, USDA
Closure Type Hinged
Capacity (English) 3.04 oz.
Volume (English) 3.04 oz.
Capacity 3.04 oz., 90mL
Quantity 50 Case
Material Polypropylene