Please login to your online account to display your discounted pricing

Dilauroyl peroxide, 99%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  349741000

 View more versions of this product

Catalog No. AC349741000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 398.63
Physical Form Powder
Quantity 100g
Color White
TSCA TSCA
RTECS Number OF2625000
EINECS Number 203-326-3
Melting Point 53° to 57°C
Synonym Dilauroyl peroxide
Health Hazard 2 GHS H Statement:
Heating may cause a fire.
Infrared Spectrum Authentic
Packaging Plastic bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/other combustible materials.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C24H46O4
Health Hazard 1 GHS Signal Word: Danger
Linear Formula [CH3(CH2)10CO]2O2
CAS 105-74-8
Assay Percent Range 99%
MDL Number MFCD00008964
Chemical Name or Material Dilauroyl peroxide
Name Note 99%

Specifications

Chemical Name or Material Dilauroyl peroxide
Name Note 99%
CAS 105-74-8
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C24H46O4
Linear Formula [CH3(CH2)10CO]2O2
Formula Weight 398.63
MDL Number MFCD00008964
Synonym Dilauroyl peroxide
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/other combustible materials.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Heating may cause a fire.

SDS & Safety Documents