Please login to your online account to display your discounted pricing

Diisopropylphosphine, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC383400500

 View more versions of this product

Catalog No. AC383400500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.
Health Hazard 2 GHS H Statement
May be fatal if swallowed and enters airways. Causes severe skin burns and eye damage. May cause damage to organs through prolonged or repeated exposure. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor.
Assay 9 to 11% (Total base)
Formula Weight 118.16
Physical Form Liquid
Quantity 50g
Color Clear yellow to pale yellow-brown
Density 0.6700g/mL
Linear Formula [(CH3)2CH]2PH
CAS 110-54-3
MDL Number MFCD00233814
CAS 20491-53-6
Packaging Glass bottle
NMR complies (31P-NMR)
Health Hazard 1 Danger
Chemical Name or Material Diisopropylphosphine
Molecular Formula C6H15P

Specifications

Chemical Name or Material Diisopropylphosphine
CAS 20491-53-6
CAS 110-54-3
Assay 9 to 11% (Total base)
Molecular Formula C6H15P
Linear Formula [(CH3)2CH]2PH
Formula Weight 118.16
MDL Number MFCD00233814
Accessories

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

GHS H Statement
May be fatal if swallowed and enters airways. Causes severe skin burns and eye damage. May cause damage to organs through prolonged or repeated exposure. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor.

SDS & Safety Documents