Please login to your online account to display your discounted pricing

Diisopropyldichlorosilane, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Diisopropyldichlorosilane
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H14Cl2Si
Formula Weight 185.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC432360250

  SDS
thermo fisher scientific
AC432360250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC432360050

  SDS
thermo fisher scientific
AC432360050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 185.17
Physical Form Liquid
Quantity 25g
Color Colorless
Density 1.0260g/mL
Refractive Index 1.4430 to 1.4450 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Contact with water liberates toxic gas.
Assay Percent Range 97%
CAS 7751-38-4
Molecular Formula C6H14Cl2Si
Chemical Name or Material Diisopropyldichlorosilane
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement:
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Contact with water liberates toxic gas.