missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diisopropylamine OR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 108-18-9
Molecular Formula ((CH3)2CH)2NH
Formula Weight 101.19
Chemical Name or Material Diisopropylamine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK23852 Avantor Performance
238562
SAFEMOR 2kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

108-18-9
((CH3)2CH)2NH
101.19
Diisopropylamine

Specifications

108-18-9
((CH3)2CH)2NH
101.19
SAFEMOR
2kg
Diisopropylamine