missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropylamine Dichloroacetate 98.0+%, TCI America™
$37.29 - $104.38
Chemical Identifiers
| CAS | 660-27-5 |
|---|---|
| Molecular Formula | C8H17Cl2NO2 |
| Molecular Weight (g/mol) | 230.13 |
| MDL Number | MFCD00868288 |
| InChI Key | ILKBHIBYKSHTKQ-UHFFFAOYSA-N |
| Synonym | diisopropylamine dichloroacetate, diisopropylammonium dichloroacetate, diisopropylamine 2,2-dichloroacetate, vasculopatina, dipromonium, cubisol, dapocel, disotat, kalodil, oxypangam |
| PubChem CID | 12617 |
| IUPAC Name | 2,2-dichloroacetic acid; bis(propan-2-yl)amine |
| SMILES | OC(=O)C(Cl)Cl.CC(C)NC(C)C |
Chemical Identifiers
| 660-27-5 | |
| 230.13 | |
| ILKBHIBYKSHTKQ-UHFFFAOYSA-N | |
| 12617 | |
| OC(=O)C(Cl)Cl.CC(C)NC(C)C |
| C8H17Cl2NO2 | |
| MFCD00868288 | |
| diisopropylamine dichloroacetate, diisopropylammonium dichloroacetate, diisopropylamine 2,2-dichloroacetate, vasculopatina, dipromonium, cubisol, dapocel, disotat, kalodil, oxypangam | |
| 2,2-dichloroacetic acid; bis(propan-2-yl)amine |
Specifications
| 660-27-5 | |
| White | |
| MFCD00868288 | |
| diisopropylamine dichloroacetate, diisopropylammonium dichloroacetate, diisopropylamine 2,2-dichloroacetate, vasculopatina, dipromonium, cubisol, dapocel, disotat, kalodil, oxypangam | |
| OC(=O)C(Cl)Cl.CC(C)NC(C)C | |
| 230.13 | |
| 230.13 | |
| Crystalline Powder |
| 122°C | |
| C8H17Cl2NO2 | |
| 1 g | |
| ILKBHIBYKSHTKQ-UHFFFAOYSA-N | |
| 2,2-dichloroacetic acid; bis(propan-2-yl)amine | |
| 12617 | |
| ≥98.0% (T) | |
| Diisopropylamine Dichloroacetate |
Safety and Handling
RTECSNumber : AG6475000
TSCA : Yes