missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diisopropylamine dichloroacetate, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6134106

 View more versions of this product

Catalog No. AAH6134106



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diisopropylamine dichloroacetate
White
[(CH3)2CH]2NH·Cl2CHCO2H
5 g
ILKBHIBYKSHTKQ-UHFFFAOYSA-N
2,2-dichloroacetic acid;N-propan-2-ylpropan-2-amine
12617
95%
660-27-5
C8H17Cl2NO2
MFCD00868288
diisopropylamine dichloroacetate, diisopropylammonium dichloroacetate, diisopropylamine 2,2-dichloroacetate, vasculopatina, dipromonium, cubisol, dapocel, disotat, kalodil, oxypangam
OC(=O)C(Cl)Cl.CC(C)NC(C)C
230.13
230.13
Crystalline Powder

Chemical Identifiers

660-27-5
230.13
ILKBHIBYKSHTKQ-UHFFFAOYSA-N
12617
OC(=O)C(Cl)Cl.CC(C)NC(C)C
C8H17Cl2NO2
MFCD00868288
diisopropylamine dichloroacetate, diisopropylammonium dichloroacetate, diisopropylamine 2,2-dichloroacetate, vasculopatina, dipromonium, cubisol, dapocel, disotat, kalodil, oxypangam
2,2-dichloroacetic acid;N-propan-2-ylpropan-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 211-538-2

RTECSNumber : AG6475000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only