missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diisopropylamine, >99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A102800F

 View more versions of this product

Catalog No. AAA102800F



Description

Description

Applications
Diisopropylamine is used as a precursor to prepare lithium diisopropylamide and sulfenamides, which are used in rubber vulcanization. It is also involved in the synthesis of N,N-Diisopropylethylamine (Hünig′s base) obtained by alkylation with diethyl sulfate. Diisopropylammonium bromide is a bromide salt of DIPA used as an inert alternative to barium titanate. It is widely used as solvent as well as an intermediate in the the synthesis of pharmaceuticals, dyes, mineral flotation agents and emulsifiers.

Solubility
Miscible with water.

Notes
Air sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and plastics.
Specifications

Specifications

Diisopropylamine
−7°C
-61°C
108-18-9
C6H15N
diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine
CC(C)NC(C)C
101.193
101.19
0.718g/mL
83°C to 84°C
2500mL
>99%
MFCD00008862
UAOMVDZJSHZZME-UHFFFAOYSA-N
N-propan-2-ylpropan-2-amine
7912

Chemical Identifiers

108-18-9
101.193
UAOMVDZJSHZZME-UHFFFAOYSA-N
7912
CC(C)NC(C)C
C6H15N
MFCD00008862
diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine
N-propan-2-ylpropan-2-amine
Safety and Handling

Safety and Handling

EINECSNumber : 203-558-5

SDS Documents