missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diisopropylamine, 99%, ACROS Organics™

Diisopropylamine, 99%, C6H15N, CAS Number-108-18-9, 2-propanamine, n-1-methylethyl, bis propan-2-yl amine, diisopropylamine, di-isopropylamine, diisopropyl amine, dipa, n-1-methylethyl-2-propanamine, n-isopropyl-1-amino-2-methylethane, n-isopropylpropan-2-amine, n,n-diisopropylamine, 1L, 04, 154

Manufacturer:  Thermo Fisher Scientific 149460025

 View more versions of this product

Catalog No. AC149460025



Description

Description

Diisopropylamine, 99%, C6H15N, CAS Number-108-18-9, 2-propanamine, n-1-methylethyl, bis propan-2-yl amine, diisopropylamine, di-isopropylamine, diisopropyl amine, dipa, n-1-methylethyl-2-propanamine, n-isopropyl-1-amino-2-methylethane, n-isopropylpropan-2-amine, n,n-diisopropylamine, 1L, 04, 154

Specifications

Specifications

Diisopropylamine
−17°C
Glass Bottle
95mbar at 20°C
Colorless
2.5L
108-18-9
C6H15N
1158
15, 3215
Solubility in water: 100g/L (20°C). Other solubilities: soluble in alcohol, soluble in most common organic solvents
CC(C)NC(C)C
101.193
101.19
99%
0.7160g/mL
Authentic
1.3910 to 1.3930
84.0°C
-61.0°C
99%
99%
[(CH3)2CH]2NH
04, 154
2-propanamine, n-1-methylethyl, bis propan-2-yl amine, di-isopropylamine, diisopropyl amine, diisopropylamine, dipa, n,n-diisopropylamine, n-1-methylethyl-2-propanamine, n-isopropyl-1-amino-2-methylethane, n-isopropylpropan-2-amine
UAOMVDZJSHZZME-UHFFFAOYSA-N
N-propan-2-ylpropan-2-amine
7912
Liquid

Chemical Identifiers

108-18-9
101.193
2-propanamine, n-1-methylethyl, bis propan-2-yl amine, di-isopropylamine, diisopropyl amine, diisopropylamine, dipa, n,n-diisopropylamine, n-1-methylethyl-2-propanamine, n-isopropyl-1-amino-2-methylethane, n-isopropylpropan-2-amine
N-propan-2-ylpropan-2-amine
C6H15N
UAOMVDZJSHZZME-UHFFFAOYSA-N
7912
CC(C)NC(C)C
Safety and Handling

Safety and Handling

GHS P Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic if inhaled.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents