Learn More
Diisopropylamine, 99%, Thermo Scientific Chemicals
Diisopropylamine, 99%, C6H15N, CAS Number-108-18-9, 2-propanamine, n-1-methylethyl, bis propan-2-yl amine, diisopropylamine, di-isopropylamine, diisopropyl amine, dipa, n-1-methylethyl-2-propanamine, n-isopropyl-1-amino-2-methylethane, n-isopropylpropan-2-amine, n,n-diisopropylamine, 1L, 04, 154
Supplier: Thermo Scientific Chemicals 149460010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diisopropylamine | |
108-18-9 | |
Colorless | |
84.0°C | |
Authentic | |
Glass Bottle | |
1.3910 to 1.3930 | |
1 L | |
04, 154 | |
15, 3215 | |
Solubility in water: 100g/L (20°C). Other solubilities: soluble in alcohol, soluble in most common organic solvents | |
CC(C)NC(C)C | |
101.19 | |
0.4 mPa.s (20°C) | |
95mbar at 20°C | |
Liquid |
99% | |
-61.0°C | |
0.7160g/mL | |
−17°C | |
99% | |
C6H15N | |
[(CH3)2CH]2NH | |
1158 | |
0.716 | |
diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine | |
UAOMVDZJSHZZME-UHFFFAOYSA-N | |
N-propan-2-ylpropan-2-amine | |
7912 | |
101.19 | |
99% |
Chemical Identifiers
108-18-9 | |
diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine | |
N-propan-2-ylpropan-2-amine |
UAOMVDZJSHZZME-UHFFFAOYSA-N | |
7912 | |
CC(C)NC(C)C |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Danger
EINECSNumber : 203-558-5
RUO – Research Use Only