missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diisopropyl sulfide, 99%, Thermo Scientific Chemicals

$67.03 - $204.53

Chemical Identifiers

CAS 625-80-9
Molecular Formula C6H14S
Molecular Weight (g/mol) 118.24
MDL Number MFCD00008895
InChI Key XYWDPYKBIRQXQS-UHFFFAOYSA-N
Synonym diisopropyl sulfide, isopropyl sulfide, propane, 2,2'-thiobis, 2,4-dimethyl-3-thiapentane, isopropyl sulphide, diisopropyl sulphide, 2,2'-thiodipropane, 2-isopropylsulfanyl propane, 2-propan-2-ylsulfanyl propane, isopropylsulfide
PubChem CID 12264
SMILES CC(C)SC(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2130209
View Documents Promotion Details
Thermo Scientific Chemicals
B2130209
10 g
Each for $67.03
 
AAB2130218
View Documents Promotion Details
Thermo Scientific Chemicals
B2130218
50 g
Each for $204.53
 
Description

Description

Used as a flavoring agents and intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavoring agents and intermediate for organic synthesis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

625-80-9
118.24
XYWDPYKBIRQXQS-UHFFFAOYSA-N
12264
C6H14S
MFCD00008895
diisopropyl sulfide, isopropyl sulfide, propane, 2,2'-thiobis, 2,4-dimethyl-3-thiapentane, isopropyl sulphide, diisopropyl sulphide, 2,2'-thiodipropane, 2-isopropylsulfanyl propane, 2-propan-2-ylsulfanyl propane, isopropylsulfide
CC(C)SC(C)C

Specifications

625-80-9
119°C to 120°C
Stench
1.4385
MFCD00008895
UN1993
diisopropyl sulfide, isopropyl sulfide, propane, 2,2'-thiobis, 2,4-dimethyl-3-thiapentane, isopropyl sulphide, diisopropyl sulphide, 2,2'-thiodipropane, 2-isopropylsulfanyl propane, 2-propan-2-ylsulfanyl propane, isopropylsulfide
XYWDPYKBIRQXQS-UHFFFAOYSA-N
2-propan-2-ylsulfanylpropane
12264
99%
0.814
7°C (44°F)
C6H14S
(CH3)2CHSCH(CH3)2
10 g
1731257
Not miscible or difficult to mix with water.
CC(C)SC(C)C
118.24
118.24
Diisopropyl sulfide
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 210-911-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only