Please login to your online account to display your discounted pricing

Diisopropyl Phosphite 98%, ACROS Organics™

$122.32 - $122.32

Chemical Identifiers

Chemical Name or Material Diisopropyl phosphite
Name Note 98%
CAS 1809-20-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H15O3P
Linear Formula [(CH3)2CHO]2P(O)H
Formula Weight 166.16
MDL Number MFCD00117905
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC115261000

  SDS
fisher scientific acros
115261000
Glass bottle 100g Each for $122.32
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 166.16
Quantity 100g
Color Clear colorless
TSCA TSCA
EINECS Number 217-317-7
RTECS Number SZ7660000
Density 0.997g/mL
Beilstein 01, 363
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 STENCH
Infrared Spectrum Authentic
Refractive Index 1.4065 to 1.4085 (20°C, 589nm)
Packaging Glass bottle
Linear Formula [(CH3)2CHO]2P(O)H
Assay Percent Range 98%
CAS 1809-20-7
Chemical Name or Material Diisopropyl phosphite
Name Note 98%
Molecular Formula C6H15O3P
MDL Number MFCD00117905
Safety and Handling

STENCH

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.