Please login to your online account to display your discounted pricing

Alfa Aesar™ Diisopropyl phenylphosphonite, 95%

$521.34 - $521.34

Chemical Identifiers

Chemical Name or Material Diisopropyl phenylphosphonite
CAS 36238-99-0
Molecular Formula C6H5P[OCH(CH3)2]2
Formula Weight 226.26
MDL Number MFCD00015024
Synonym Phenylphosphonous acid diisopropyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2370218

  SDS
alfa aesar™
B2370218
50g Each for $521.34
Description & Specifications

Specifications

Formula Weight 226.26
Chemical Name or Material Diisopropyl phenylphosphonite
Molecular Formula C6H5P[OCH(CH3)2]2
CAS 36238-99-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 50g
MDL Number MFCD00015024
Assay 95%
Boiling Point 112° to 114°C (8mmHg)
Synonym Phenylphosphonous acid diisopropyl ester
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.