Please login to your online account to display your discounted pricing

Alfa Aesar™ (+)-Diisopropyl O,O'-bis(trimethylsilyl)-L-tartrate, 99%

$738.82 - $738.82

Chemical Identifiers

Chemical Name or Material (+)-Diisopropyl O,O'-bis(trimethylsilyl)-L-tartrate
CAS 130678-42-1
Molecular Formula C16H34O6Si2
Formula Weight 378.61g/mol
MDL Number MFCD00192055
Beilstein 3620696
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5106806

 
alfa aesar™
H51068-06
Each for $738.82
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material (+)-Diisopropyl O,O'-bis(trimethylsilyl)-L-tartrate
CAS 130678-42-1
Assay 99%
Molecular Formula C16H34O6Si2
Formula Weight 378.61g/mol
MDL Number MFCD00192055
Beilstein 3620696