Please login to your online account to display your discounted pricing

(+)-Diisopropyl L-tartrate, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  212030250

 View more versions of this product

Catalog No. AC212030250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Viscous Liquid
Formula Weight 234.25
Quantity 25g
Color Clear colorless
Beilstein 03, 517
EINECS Number 218-709-0
TSCA TSCA
Specific Rotation Condition +17.25° (20°C neat)
Density 1.11g/mL
RTECS Number WW8100000
Fieser 13,276; 16,339; 12,493; 13,310
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.4380 to 1.4400 (20°C, 589nm)
Health Hazard 1 HYGROSCOPIC
Chemical Name or Material (+)-Diisopropyl L-tartrate
Specific Rotation +16° to +18.5° (20°C, 589nm) (neat)
Name Note 99%
Assay Percent Range 99%
Molecular Formula C10H18O6
Linear Formula {-CH(OH)CO2CH(CH3)2}2
MDL Number MFCD00064450
CAS 2217-15-4
CAS Min % 98.5
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (+)-Diisopropyl L-tartrate
Name Note 99%
CAS 2217-15-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H18O6
Linear Formula {-CH(OH)CO2CH(CH3)2}2
Formula Weight 234.25
MDL Number MFCD00064450
Accessories

HYGROSCOPIC

SDS & Safety Documents