Please login to your online account to display your discounted pricing

(+)-Diisopropyl L-tartrate, 99%, ACROS Organics™

$33.21 - $33.21

Chemical Identifiers

Chemical Name or Material (+)-Diisopropyl L-tartrate
Name Note 99%
CAS 2217-15-4
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C10H18O6
Linear Formula {-CH(OH)CO2CH(CH3)2}2
Formula Weight 234.25g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC212030250

  SDS
fisher scientific acros
212030250
Glass bottle 25g Each for $33.21
Description & Specifications

Specifications

Density 1.11g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4380 to 1.4400 (20°C, 589nm)
Specific Rotation +16° to +18.5° (20°C, 589nm) (neat)
Specific Rotation Condition +17.25° (20°C neat)
Color Clear colorless
Quantity 25g
Chemical Name or Material (+)-Diisopropyl L-tartrate
Name Note 99%
CAS 2217-15-4
CAS Min % 98.5
CAS Max % 100.0
Physical Form Clear colorless viscous liquid
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C10H18O6
Linear Formula {-CH(OH)CO2CH(CH3)2}2
Formula Weight 234.25g/mol
MDL Number MFCD00064450
Beilstein 03, 517
Fieser 13,276; 16,339; 12,493; 13,310
Health Hazard 1 HYGROSCOPIC
RTECS Number WW8100000
EINECS Number 218-709-0
TSCA TSCA