missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropyl Fumarate 98.0+%, TCI America™
$63.38 - $212.51
Chemical Identifiers
| CAS | 7283-70-7 |
|---|---|
| Molecular Formula | C10H16O4 |
| Molecular Weight (g/mol) | 200.23 |
| MDL Number | MFCD00545172 |
| InChI Key | FNMTVMWFISHPEV-AATRIKPKSA-N |
| Synonym | Fumaric Acid Diisopropyl Ester |
| PubChem CID | 5357078 |
| IUPAC Name | 1,4-bis(propan-2-yl) (2E)-but-2-enedioate |
| SMILES | CC(C)OC(=O)\C=C\C(=O)OC(C)C |
Chemical Identifiers
| 7283-70-7 | |
| 200.23 | |
| FNMTVMWFISHPEV-AATRIKPKSA-N | |
| 5357078 | |
| CC(C)OC(=O)\C=C\C(=O)OC(C)C |
| C10H16O4 | |
| MFCD00545172 | |
| Fumaric Acid Diisopropyl Ester | |
| 1,4-bis(propan-2-yl) (2E)-but-2-enedioate |
Specifications
| 7283-70-7 | |
| Colorless | |
| C10H16O4 | |
| 25 g | |
| FNMTVMWFISHPEV-AATRIKPKSA-N | |
| 1,4-bis(propan-2-yl) (2E)-but-2-enedioate | |
| 5357078 | |
| ≥98.0% (GC) | |
| Diisopropyl Fumarate |
| 2°C | |
| 110°C | |
| MFCD00545172 | |
| Fumaric Acid Diisopropyl Ester | |
| CC(C)OC(=O)\C=C\C(=O)OC(C)C | |
| 200.23 | |
| 200.23 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1099&(2)-1142
RTECSNumber : LT1575000
TSCA : No