missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Riedel-de Haen™ Diisopropyl ether, ACS Reagent, contains either BHT or hydroquinone as stabilizer, ≥99.0%, Honeywell Riedel-de Haën™

ACS Reagent, contains either BHT or hydroquinone as stabilizer,≥99.0%

$862.15 - $862.15

Chemical Identifiers

CAS 108-20-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00008880
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046769 View Documents Honeywell Riedel-de Haen™
3982766X1L
6 x 1L Fiberboard box Case of 6 Each for $862.15
 
Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Chemical Identifiers

108-20-3
102.177
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
7914
CC(C)OC(C)C
C6H14O
MFCD00008880
diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
2-propan-2-yloxypropane

Specifications

0.725g/mL at 25°C
Odorless
3.5 (vs air)
Undesignated
6 x 1L
C6H14O
MFCD00008880
1731256
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
2-propan-2-yloxypropane
7914
≥99%
−29°C (−20.2°F)
Fiberboard box
67°C to 70°C (1013hPa)
-86°C
108-20-3
(CH3)2CHOCH(CH3)2
UN1159
diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
CC(C)OC(C)C
102.177
102.17g/mol
Diisopropyl ether
Safety and Handling

Safety and Handling

P210-P273-P403 + P235

DOTInformation : class 3 - PG 2 - Diisopropyl ether

RTECSNumber : 203-560-6