Please login to your online account to display your discounted pricing

Alfa Aesar™ (-)-Diisopropyl D-tartrate, 98%

$82.35 - $303.24

Chemical Identifiers

Chemical Name or Material (-)-Diisopropyl D-tartrate
CAS 62961-64-2
Molecular Formula C10H18O6
Formula Weight 234.25
MDL Number MFCD00008876
Beilstein 5265431
Synonym D-Tartaric acid diisopropyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1736214

  SDS
alfa aesar™
A1736214
25g Each for $82.35

AAA1736222

  SDS
alfa aesar™
A1736222
100g Each for $303.24
Description & Specifications

Specifications

Formula Weight 234.25
Chemical Name or Material (-)-Diisopropyl D-tartrate
TSCA TSCA
Assay 98%
Boiling Point 150° to 151°C (11mmHg)
Molecular Formula C10H18O6
Quantity 25g
Flash Point 110°C (230°F)
MDL Number MFCD00008876
Optical Rotation −16° (Neat)
CAS 62961-64-2
Beilstein 5265431
Synonym D-Tartaric acid diisopropyl ester