Learn More
Diisopropyl bromomethylphosphonate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1376314
Description
Diisopropyl bromomethylphosphonate, is used as an organophosphorus intermediate. It is also used as a biochemical reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiisopropyl bromomethylphosphonate, is used as an organophosphorus intermediate. It is also used as a biochemical reagent.
Solubility
Insoluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Diisopropyl bromomethylphosphonate | |
80°C (1 mmHg) | |
1.4495 | |
25 g | |
4740388 | |
Insoluble in water. | |
CC(C)OP(=O)(CBr)OC(C)C | |
259.08 | |
259.09 |
98432-80-5 | |
C7H16BrO3P | |
MFCD00236036 | |
UN3265 | |
diisopropyl bromomethylphosphonate, bromomethylphosphonic acid diisopropyl ester, diisopropyl bromomethyl phosphonate, bromomethylphosphonate diisopropyl ester, 2-bromomethyl propan-2-yloxy phosphoryl oxypropane | |
JNONGIUWXHOEEN-UHFFFAOYSA-N | |
2-[bromomethyl(propan-2-yloxy)phosphoryl]oxypropane | |
4366885 | |
97% |
Chemical Identifiers
98432-80-5 | |
259.08 | |
JNONGIUWXHOEEN-UHFFFAOYSA-N | |
4366885 | |
CC(C)OP(=O)(CBr)OC(C)C |
C7H16BrO3P | |
MFCD00236036 | |
diisopropyl bromomethylphosphonate, bromomethylphosphonic acid diisopropyl ester, diisopropyl bromomethyl phosphonate, bromomethylphosphonate diisopropyl ester, 2-bromomethyl propan-2-yloxy phosphoryl oxypropane | |
2-[bromomethyl(propan-2-yloxy)phosphoryl]oxypropane |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only