Please login to your online account to display your discounted pricing

(+)-Diisopinocampheylchloroborane, 1.8M solution in hexanes, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC388421000

 View more versions of this product

Catalog No. AC388421000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
May be fatal if swallowed and enters airways. Suspected of damaging fertility. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement
Do NOT induce vomiting. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection.
Formula Weight 320.77
Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can
Appearance of Solution Clear solution
Molecular Formula C20H34BCl
CAS 64742-89-8
MDL Number MFCD00074808
CAS 112246-73-8
CAS 80-56-8
CAS 80-56-8
CAS 71-43-2
Chemical Name or Material (+)-Diisopinocampheylchloroborane
Health Hazard 1 Danger

Specifications

Chemical Name or Material (+)-Diisopinocampheylchloroborane
CAS 112246-73-8
CAS 80-56-8
CAS 80-56-8
CAS 71-43-2
CAS 64742-89-8
Molecular Formula C20H34BCl
Formula Weight 320.77
MDL Number MFCD00074808
Accessories

Danger

GHS P Statement
Do NOT induce vomiting. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection.

GHS H Statement
May be fatal if swallowed and enters airways. Suspected of damaging fertility. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor.

SDS & Safety Documents