Please login to your online account to display your discounted pricing

Alfa Aesar™ Diisopentylamine, 97%


Quantity 5g

Manufacturer: alfa aesar™  L0229706

 View more versions of this product

Catalog No. AAL0229706

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 157.3
CAS 544-00-3
MDL Number MFCD00015053
Molecular Formula [(CH3)2CHCH2CH2]2NH
Synonym Bis(3-methylbutyl)amine; Diisoamylamine
Chemical Name or Material Diisopentylamine
TSCA TSCA
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Boiling Point 187° to 188°C
Melting Point −44°C
Quantity 5g
Merck Index 14,3193
UN Number UN2733
Flash Point 58°C (136°F)
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Beilstein 1719663
Health Hazard 1 Danger

Specifications

Chemical Name or Material Diisopentylamine
CAS 544-00-3
Molecular Formula [(CH3)2CHCH2CH2]2NH
Formula Weight 157.3
MDL Number MFCD00015053
UN Number UN2733
Beilstein 1719663
Merck Index 14,3193
Synonym Bis(3-methylbutyl)amine; Diisoamylamine

Applications
Intermediate for organic synthesis

Solubility
Soluble in ethanol, ethyl ether and chloroform. Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Accessories

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

SDS & Safety Documents