Please login to your online account to display your discounted pricing

Alfa Aesar™ Diisopentyl sulfide, 98%

$38.18 - $38.18

Chemical Identifiers

Chemical Name or Material Diisopentyl sulfide
CAS 544-02-5
Molecular Formula [(CH3)2CHCH2CH2]2S
Formula Weight 174.35
MDL Number MFCD00026522
Synonym Bis(3-methylbutyl) sulfide; Isopentyl sulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2303006

 
alfa aesar™
B2303006
5g Each for $38.18
Description & Specifications

Specifications

Formula Weight 174.35
Chemical Name or Material Diisopentyl sulfide
TSCA TSCA
Assay 98%
Boiling Point 79° to 80°C (7mmHg)
Molecular Formula [(CH3)2CHCH2CH2]2S
Quantity 5g
Odor Unpleasant
MDL Number MFCD00026522
CAS 544-02-5
Synonym Bis(3-methylbutyl) sulfide; Isopentyl sulfide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Diisopentyl sulfide is used as chemical, organic intermediates.

Solubility
Soluble in water (5.67mg/L @ 25°C.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.