missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diisopentyl ether, mixture of isomers, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A11231AP
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diisopentyl ether | |
544-01-4 | |
172°C to 174°C | |
C10H22O | |
MFCD00008947 | |
UN3271 | |
14,5118 | |
AQZGPSLYZOOYQP-UHFFFAOYSA-N | |
3-methyl-1-(3-methylbutoxy)butane | |
10989 | |
96% |
Mixture of isomers | |
0.78 | |
46°C (115°F) | |
1.407 | |
500 mL | |
1698014 | |
diisopentyl ether, isoamyl ether, isopentyl ether, diisoamyl ether, isoamyl oxide, butane, 1,1'-oxybis 3-methyl, di-3-methylbutyl ether, di-iso-amyl ether, 1,1'-oxybis 3-methylbutane, 1,1'-oxylbis 3-methylbutane | |
CC(C)CCOCCC(C)C | |
158.285 | |
158.29 |
Chemical Identifiers
544-01-4 | |
158.285 | |
AQZGPSLYZOOYQP-UHFFFAOYSA-N | |
10989 | |
CC(C)CCOCCC(C)C |
C10H22O | |
MFCD00008947 | |
diisopentyl ether, isoamyl ether, isopentyl ether, diisoamyl ether, isoamyl oxide, butane, 1,1'-oxybis 3-methyl, di-3-methylbutyl ether, di-iso-amyl ether, 1,1'-oxybis 3-methylbutane, 1,1'-oxylbis 3-methylbutane | |
3-methyl-1-(3-methylbutoxy)butane |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber : 208-857-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only