Please login to your online account to display your discounted pricing

Alfa Aesar™ Diisobutylaluminum hydride, 1M solution in toluene, packaged under Argon in resealable ChemSeal™ bottles

$81.68 - $81.68

Chemical Identifiers

Chemical Name or Material Diisobutylaluminum hydride
CAS 1191-15-7
Linear Formula [(CH3)2CHCH2]2AlH
Formula Weight 142.22
MDL Number MFCD00008928
UN Number UN3399
Beilstein 4123663
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42591AD

  SDS
alfa aesar™
42591AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42591AH

  SDS
alfa aesar™
42591AH
200mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42591AP

  SDS
alfa aesar™
42591AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42591AE

  SDS
alfa aesar™
42591AE
100mL Each for $81.68
Description & Specifications

Specifications

Formula Weight 142.22
Health Hazard 2 GHS H Statement
H224-H260-H304-H361-H373-H314-H318-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs (state all organs affected, if known) through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Liquid
Density 0.805g/mL
Packaging ChemSeal™ bottles
Chemical Name or Material Diisobutylaluminum hydride
Concentration 1M solution in toluene
Sensitivity Air & Moisture Sensitive
TSCA TSCA
Boiling Point ≈110°C
Linear Formula [(CH3)2CHCH2]2AlH
MDL Number MFCD00008928
CAS 1191-15-7
Quantity 50mL
UN Number UN3399
Beilstein 4123663
Flash Point 4°C (40°F)
Health Hazard 1 Danger
Color Colorless
Storage Note 1
  • Packaged under Argon
DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

  • Packaged under Argon

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H224-H260-H304-H361-H373-H314-H318-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs (state all organs affected, if known) through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause drowsiness or dizziness.

  • Packaged under Argon