missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diisobutylaluminium hydride, 1.2M (20 wt%) solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 201084000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid, Reduction reagentSpecifications
Diisobutylaluminium hydride | |
0.8480g/mL | |
4°C | |
C8H19Al | |
MFCD00008928 | |
04, IV, 4400 | |
0.848 | |
DIBAL-H | |
142.22 | |
Solution |
1191-15-7,108-88-3 | |
110.0°C | |
AcroSeal™ Glass Bottle | |
[(CH3)2CHCH2]2AIH | |
400 mL | |
01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123 | |
15, 3212 | |
Solubility in water: reacts violently. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons | |
142.22 |
RUO – Research Use Only