missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diisobutylaluminium hydride, 1M solution in hexane, AcroSeal™, ACROS Organics™

Manufacturer:  Acros Organics 183794000

 View more versions of this product

Catalog No. AC183794000



Description

Description

Lewis acid, Reduction reagent
Specifications

Specifications

Diisobutylaluminium hydride
−23°C
Undesignated
400mL
1191-15-7,110-54-3
[(CH3)2CHCH2]2AlH
04, IV, 4400
15, 3212
142.22
0.7010g/mL
AcroSeal™ Glass Bottle
-70.0°C
1M Solution in Hexane
C8H19Al
MFCD00008928
01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123
DIBAL-H
Solution

Chemical Identifiers

1191-15-7
C8H19Al
MFCD00008928
DIBAL-H
Safety and Handling

Safety and Handling

GHS H Statement
May be fatal if swallowed and enters airways. Toxic to aquatic life with long lasting effects. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure. Suspected of damaging fertility. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Catches fire spontaneously if exposed to air. Reacts violently with water.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Handle under inert gas. Protect from moisture. IF ON SKIN: Immerse in cool water/wrap in wet bandages. Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Avoid release to the environment. Store in a dry place. Store in a closed container. Do not allow contact with air.

Danger

SDS Documents