missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diisobutylaluminium hydride, 1.0M solution in heptane, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC201058000

 View more versions of this product

Catalog No. AC201058000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Solution
800mL
AcroSeal™ Glass Bottle
04, IV, 4400
01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123
142-82-5,1191-15-7
1.0M Solution in Heptane
MFCD00008928
Diisobutylaluminium hydride
142.22
Clear
0.7000g/mL
15, 3212
DIBAL-H, 1.0M solution in heptane
19.5 to 20.5% (DIBAL-H, on Al basis)
C8H19Al
[(CH3)2CHCH2]2AIH

Chemical Identifiers

1191-15-7,142-82-5
[(CH3)2CHCH2]2AIH
MFCD00008928
Diisobutylaluminium hydride, 1.0M Solution in Heptane
C8H19Al
142.22
DIBAL-H, 1.0M solution in heptane

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger