Learn More
Diisobutylaluminium chloride, 0.8M solution in heptane, AcroSeal™, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 377568000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Lewis acidSpecifications
Diisobutylaluminium chloride | |
0.7260g/mL | |
AcroSeal™ Glass bottle | |
MFCD00008927 | |
0.726 | |
Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons | |
CC(C)C[Al](CC(C)C)Cl | |
176.67 | |
176.67 |
1779-25-5,142-82-5 | |
−4°C | |
C8H18AlCl | |
800mL | |
diisobutylaluminum chloride, chlorodiisobutylaluminium, chlorodiisobutylaluminum, diisobutylchloroaluminum, aluminum, chlorobis 2-methylpropyl, aluminum, chlorodiisobutyl, bis isobutyl aluminum chloride, diisobutylaluminum monochloride, chlorobis 2-methylpropyl aluminum, alluminio diisobutil-monocloruro italian | |
HQMRIBYCTLBDAK-UHFFFAOYSA-M | |
chloro-bis(2-methylpropyl)alumane | |
15688 |
Chemical Identifiers
1779-25-5 | |
176.67 | |
HQMRIBYCTLBDAK-UHFFFAOYSA-M | |
15688 | |
CC(C)C[Al](CC(C)C)Cl |
C8H18AlCl | |
MFCD00008927 | |
diisobutylaluminum chloride, chlorodiisobutylaluminium, chlorodiisobutylaluminum, diisobutylchloroaluminum, aluminum, chlorobis 2-methylpropyl, aluminum, chlorodiisobutyl, bis isobutyl aluminum chloride, diisobutylaluminum monochloride, chlorobis 2-methylpropyl aluminum, alluminio diisobutil-monocloruro italian | |
chloro-bis(2-methylpropyl)alumane |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
In contact with water releases
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Im
GHS Signal Word: Danger
EINECSNumber : 217-216-8
RUO â Research Use Only