missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diisobutyl methylphosphonate, 97%

Manufacturer:  Alfa Aesar™ 4464304

 View more versions of this product

Catalog No. AA4464304



Specifications

Specifications

Diisobutyl methylphosphonate
7242-56-0
MFCD00015638
IGNFYWATZHBYFJ-UHFFFAOYSA-N
2-methyl-1-[methyl(2-methylpropoxy)phosphoryl]oxypropane
566101
Liquid
2g
C9H21O3P
1-isobutoxy methyl phosphoryl oxy-2-methyl-propane
CC(C)COP(=O)(C)OCC(C)C
208.238
208.24
97%

Chemical Identifiers

7242-56-0
208.238
IGNFYWATZHBYFJ-UHFFFAOYSA-N
566101
CC(C)COP(=O)(C)OCC(C)C
C9H21O3P
MFCD00015638
1-isobutoxy methyl phosphoryl oxy-2-methyl-propane
2-methyl-1-[methyl(2-methylpropoxy)phosphoryl]oxypropane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents