Please login to your online account to display your discounted pricing

Alfa Aesar™ Diisobutyl methylphosphonate, 97%

$1,066.78 - $1,066.78

Chemical Identifiers

Chemical Name or Material Diisobutyl methylphosphonate
CAS 7242-56-0
Molecular Formula C9H21O3P
Formula Weight 208.24
MDL Number MFCD00015638
Synonym Methylphosphonic acid diisobutyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4464309

  SDS
alfa aesar™
4464309
10g Each for $1,066.78
Description & Specifications

Specifications

Formula Weight 208.24
Physical Form Liquid
Quantity 10g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
MDL Number MFCD00015638
Molecular Formula C9H21O3P
CAS 7242-56-0
Chemical Name or Material Diisobutyl methylphosphonate
Assay 97%
Synonym Methylphosphonic acid diisobutyl ester
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.