Please login to your online account to display your discounted pricing

Alfa Aesar™ Diisobutyl adipate, 97%

$40.82 - $509.28

Chemical Identifiers

Chemical Name or Material Diisobutyl adipate
CAS 141-04-8
Molecular Formula (CH3)2CHCH2O2C(CH2)4CO2CH2CH(CH3)2
Formula Weight 258.36
MDL Number MFCD00053722
Synonym Adipic acid diisobutyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1918614

  SDS
alfa aesar™
A1918614
25g Each for $40.82

AAA1918622

  SDS
alfa aesar™
A1918622
100g Each for $122.95

AAA1918636

  SDS
alfa aesar™
A1918636
500g Each for $509.28
Description & Specifications

Specifications

Formula Weight 258.36
Boiling Point 293°C
Molecular Formula (CH3)2CHCH2O2C(CH2)4CO2CH2CH(CH3)2
Quantity 25g
Synonym Adipic acid diisobutyl ester
Flash Point >110°C (230°F)
Chemical Name or Material Diisobutyl adipate
Assay 97%
RTECS Number AV1480000
TSCA TSCA
MDL Number MFCD00053722
CAS 141-04-8