Please login to your online account to display your discounted pricing

Diiodosilane, ACROS Organics™

$42.28 - $42.28

Chemical Identifiers

Chemical Name or Material Diiodosilane
CAS 13760-02-6
Molecular Formula H2I2Si
Linear Formula SiH2(I)2
Formula Weight 283.90g/mol
MDL Number MFCD00075171
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC388920010

  SDS
thermo fisher scientific
388920010
Glass bottle 1g Each for $42.28
Description & Specifications

Specifications

Density 2.8340g/mL
NMR Complies
Packaging Glass bottle
Color Clear
Quantity 1g
Chemical Name or Material Diiodosilane
CAS 13760-02-6
Physical Form Clear liquid
Molecular Formula H2I2Si
Linear Formula SiH2(I)2
Formula Weight 283.90g/mol
MDL Number MFCD00075171
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Reduction reagent

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Reacts violently with water.